Learn More
Thermo Scientific™ Remel™ Anaerobic Reducible LKV Blood Agar/ PEA w/Sheep Blood
Shop All Thermo Scientific ProductsSpecifications
Specifications
| Certifications/Compliance | USDA, APHIS |
| Form | Solid |
| Manufacturing Origin | United States |
| Media Type | Anaerobic reducible LKV Blood Agar/PEA with Sheep Blood |
| Formulation | Kanamycin Sulphate <1.0 |
| For Use With (Application) | Isolation and differentiation of microorganisms from samples or specimens |
| Packaging Type | Biplate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.