Learn More
Thermo Scientific™ Remel™ Bacillus paranthracis ATCC™ 61™, Culti-Loops™
Catalog No. R4601216 Shop All Thermo Scientific Products
Change view
Click to view available options
Bacillus paranthracis ATCC 61
Catalog No.
R4601216 Bacillus paranthracis ATCC 61
1 options
This item is not returnable. View return policy
Catalog No. R4601216 Supplier Thermo Scientific™ Supplier No. R4601216
Add to Cart
Add to Cart
This item is not returnable. View return policy
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
Certifications
Culti-Loops are CE marked.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 61™*
Product Type Culti-Loop
Strain Designations AMC 732 [LMG 16633, NRS 727, Vitek #200284]
Species Bacillus paranthracis
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.