Learn More
Thermo Scientific™ Remel™ Bacteroides fragilis ATCC™ 23745™, Culti-Loops™
Shop All Thermo Scientific ProductsDescription
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
Specifications
Specifications
| ATCC® | 23745™* |
| Product Type | Culti-Loop |
| Strain Designations | WDCM00055, [ICPB 3498, NCTC l0581] |
| Species | Bacteroides fragilis |
Culti-Loops are CE marked.