Learn More
Thermo Scientific™ Remel™ BactiSwab™ Liquid Collection and Transport Systems
Shop All Thermo Scientific ProductsDescription
Rayon-tipped, plastic-shaft swabs are mounted in the cap of a translucent plastic tube with spiral chamber. No transport media.
Available in several formats:
- Single swabs with single plastic shaft, white cap
- Double swabs with double plastic shaft, red cap
- Liquid Stuarts, Wire Shaft, with mini-tip swab suitable for ear, nose, throat, eye, male urethral, and pediatric specimen collection; straight aluminum-shaft; green cap
Specifications
Specifications
| Product Type | Transport Swab |
| For Use With (Application) | Provides excellent recovery in routine bacteriology and is compatible with many rapid screening methods |
| Quantity | 1000/Cs. |
| Shaft Material | Plastic, Stainless Steel |