Learn More
Thermo Scientific™ Bile Esculin Azide Agar with Vancomycin

Description
Isolate and presumptively identify vancomycin-resistant enterococci (VRE) from surveillance cultures with Thermo Scientific™ Remel™ Bile Esculin Azide Agar with Vancomycin. Sodium azide and 1% oxgall are inhibitory to Gram-negative bacteria other than group D streptococci. Vancomycin (6μg/mL) is added to select for resistant strains of enterococci. Differentiation is facilitated by the hydrolysis of esculin in the presence of bile, resulting in the production of esculetin and dextrose. Ferric ammonium citrate supplies ferric ions which react with esculitin to form a black-brown complex.
Members of the genus Enterococcus are normal residents of the gastrointestinal and biliary tracts1,2. Enterococci have become significant agents of human disease, largely because of their evolving resistance to antimicrobial agents. VRE surveillance-screening has been found to facilitate earlier identification of colonized patients leading to more efficient containment of the organism. Timely identification of VRE is improved with the use of the primary isolation medium for surveillance cultures.
Sahm et al. demonstrated that Bile Esculin Azide Agar with 6μg/mL Vancomycin provides for rapid presumptive identification of VRE3.
- Selective: selective agents sodium azide and 1% oxgall are inhibitory to Gram-negative bacilli and most Gram-positive bacteria other than group D streptococci while vancomycin (6μg/mL) selects for resistant strains of enterococci.
- Easy-to-read: vancomycin-resistant enterococci are easy to read due to brown-black complex surrounding the colonies
- Ready-to-use: convenience of prepared media.
Esculin in the medium allows for early recognition of presumptive enterococcal isolates, selective agents inhibit the growth of most commensal microbial flora, and vancomycin provides additional selectivity against vancomycin-sensitive and other Gram-positive organisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Bile Esculin Azide Agar w/Vancomycin |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.