Learn More
Thermo Scientific™ Remel™ Blood Agar/ Blood Agar (TSA w/5% Sheep Blood)
Shop All Thermo Scientific ProductsSpecifications
Specifications
| Certifications/Compliance | USDA, APHIS |
| Manufacturing Origin | United States |
| Media Type | Blood Agar/Blood Agar (TSA with 5% Sheep Blood) |
| For Use With (Application) | Isolation and differentiation of microorganisms from samples or specimens |
| Packaging Type | Biplate |
| Quantity | 100/Pk. |