Learn More
Thermo Scientific™ Remel™ Blood Agar (TSA w/5% Sheep Blood)/ EMB Agar, Levine
GSA_VA
Catalog No. R02040 Shop All Thermo Scientific Products
Change view
Click to view available options
Blood Agar (TSA 5% Sheep Blood) // EMB Agar (Levine); Format: Biplate; pack of 10
Catalog No.
R02040 Blood Agar (TSA 5% Sheep Blood) // EMB Agar (Levine); Format: Biplate; pack of 10
1 options
This item is not returnable. View return policy
Catalog No. R02040 Supplier Thermo Scientific™ Supplier No. R02040
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Used for the isolation and/or differentiation of microorganisms or specimens
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USDA, APHIS
Manufacturing Origin United States
Media Type Blood Agar (TSA with 5% Sheep Blood)/EMB Agar, Levine
For Use With (Application) Isolation and differentiation of microorganisms from samples or specimens
Packaging Type Biplate
Quantity 10/Pk.