Learn More
Thermo Scientific™ Remel™ Bordetella bronchiseptica ATCC™ 4617, Culti-Loops™
Catalog No. R4601223 Shop All Thermo Scientific Products
Change view
Click to view available options
Species:
Bordetella bronchiseptica
Catalog No. Species
R4601223 Bordetella bronchiseptica
1 options
This item is not returnable. View return policy
Catalog No. R4601223 Supplier Thermo Scientific™ Supplier No. R4601223
Add to Cart
Add to Cart
This item is not returnable. View return policy
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 4617™*
Product Type Culti-Loop
Strain Designations NRRL B-140
Species Bordetella bronchiseptica
Culti-Loops are CE marked.