Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Lactose Broth

Catalog No. r112406 Shop All Fisherbrand Products
Change view
Click to view available options
Appearance:
Light to Medium Amber, Clear Broth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Appearance
R112406 Light to Medium Amber, Clear Broth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R112406
This item is not returnable. View return policy

For detection of coliform bacteria in water, foods, and dairy products according to standard methods

Compliance

Industrial Reference: AOAC, BAM, COMPF, EPA, USDA, USP.

TRUSTED_SUSTAINABILITY

Specifications

Appearance Light to Medium Amber, Clear Broth
For Use With (Application) Detection of coliform bacteria in water, foods, and dairy products according to standard methods
No. per Pack 10/Pk.
Quantity 225 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.