Learn More
Thermo Scientific™ Remel™ MacConkey Agar w/1μg/mL Ciprofloxacin
Catalog No. R01545 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
R01545 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R01545 Supplier Thermo Scientific™ Supplier No. R01545
Add to Cart
Add to Cart
This item is not returnable. View return policy
A solid medium recommended for use in qualitative procedures for primary, selective isolation of fluoroquinolone-resistant enteric gram-negative bacilli in clinical specimens
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10/Pk.