Learn More
Thermo Scientific™ Remel™ Parabacteroides distasonis ATCC™ 8503, Culti-Loops™
Catalog No. R4601226 Shop All Thermo Scientific Products
Change view
Click to view available options
Species:
Parabacteroides distasonis
Catalog No. Species
R4601226 Parabacteroides distasonis
1 options
This item is not returnable. View return policy
Catalog No. R4601226 Supplier Thermo Scientific™ Supplier No. R4601226
Add to Cart
Add to Cart
This item is not returnable. View return policy
Perform testing of media, stains, reagents, and identification kits, and evaluate bacteriological procedures with Thermo Scientific ™ Remel™ Parabacteroides distasonis ATCC™ 8503, Culti-Loops™.
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 8503™*
Product Type Culti-Loops Quality Control Organism
Species Parabacteroides distasonis
Culti-Loops are CE marked.