Learn More
Thermo Scientific™ Remel™ Phosphate Buffer w/Polysorbate 80 (4%)
Shop All Thermo Scientific ProductsDescription
Compliance
Industrial Reference: USP.
Specifications
Specifications
| Form | Liquid |
| Manufacturing Origin | United States |
| Fill Volume | 90 mL |
| For Use With (Application) | For use as a diluent in microbial limit testing |
| Packaging Type | Bottle |
| Quantity | 90 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.