Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel Prepared Media: Jones Kendric Transport to Lowenstein Jensen Mycobactocel

Catalog No. R112402 Shop All Thermo Scientific Products
Change view
Click to view available options
Lactose Broth; Format: Wide Mouth Bottle w/screw cap; Size: 90mL; pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
R112402 Lactose Broth; Format: Wide Mouth Bottle w/screw cap; Size: 90mL; pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R112402 Supplier Thermo Scientific™ Supplier No. R112402

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Form Liquid
Manufacturing Origin United States
Media Type Lactose broth
For Use With (Application) Detection of Coliform bacteria in Water, foods and dairy products according to standard method
Packaging Type Wide Mouth Bottle with Screw Cap
Quantity 90 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.