Learn More
Thermo Scientific™ Remel™ Sterile Airway Venting and Subculture Needle Units
Catalog No. R223766 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
Catalog No. Quantity
R223766 100/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R223766 Supplier Thermo Scientific™ Supplier No. R223766
Add to Cart
Add to Cart
This item is not returnable. View return policy
Needle Unit for venting aerobic blood culture bottles and subculturing blood culture bottles
  • For venting aerobic blood culture bottles and subculturing blood culture bottles
TRUSTED_SUSTAINABILITY
Specifications
Product Type Airway Needle
Sterility Sterile
For Use With (Application) For venting aerobic Blood culture bottles and subculturing Blood culture bottles
Quantity 100/Pk.