Learn More
Epredia™ Richard-Allan Scientific™ Cyto-Stain™ G
Catalog No. 23900654 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
23-900-654 1 gal.
1 options
Catalog No. 23-900-654 Supplier Epredia™ Supplier No. 7501G
Add to Cart
Add to Cart
Produces results similar to those achieved with conventional Papanicolaou stains
  • Can be used for all gynecological and non-gynecological procedures
  • Designed to produce greener cyanophilic hues in intermediate and basal cells
  • Requires one counterstain instead of two
  • Reduces staining time
Specifications
Quantity 1 gal.
Product Type Cytostain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov