Learn More
Epredia™ Dry Specimen Containers - Screw-top
Catalog No. 22046338 Shop All Epredia Products
Change view
Click to view available options
Capacity (Metric):
60 mL
120 mL
180 mL
480 mL
Material:
Polypropylene
Polystyrene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material
22-046338 480 mL Polypropylene
22-046-330 60 mL Polystyrene
22-046334 120 mL Polystyrene
22-046336 180 mL Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-046338 Supplier Epredia™ Supplier No. 534802
Add to Cart
Add to Cart
Dry specimen containers constructed of clear and opaque polystyrene, polypropylene or polyethylene with a variety of lid options.
Specifications
Product Type Dry Specimen Container
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 480 mL
Closure Type Screw Top
Color Clear
No. per Case 60/Cs.