Learn More
Epredia™ Envi-Ro-Tech™ Freezing Spray
Catalog No. 6769038 Shop All Epredia Products
Change view
Click to view available options
Envi-Ro-Tech Freezer Spray
Catalog No.
6769038 Envi-Ro-Tech Freezer Spray
1 options
Catalog No. 6769038 Supplier Epredia™ Supplier No. 6769038
Ozone-friendly freezing spray for use at the microtome and cryostat
  • Facilitates rapid freezing and consistent results
  • Attachable capillary nozzle enables spot freezing
  • Contains 1,1,1,2 Tetrafluoroethane
Specifications
Description 8 oz, can; 6/Cs
Product Type Freezing Spray