Learn More
Epredia™ Trak 300™ Oil for the HEMATRAK™ system
GSA_VA
Catalog No. 22110680 Shop All Epredia Products
Change view
Click to view available options
Quantity:
16 oz.
Catalog No. Quantity
22-110-680 16 oz.
1 options
This item is not returnable. View return policy
Catalog No. 22-110-680 Supplier Epredia™ Supplier No. M8000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Intended for use with the HEMATRAK™ automated differential system
  • Oil does not contain PCBs and is safe to use with valuable optics
  • Index of refraction is 1.5150 and viscosity is 300 cs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Application) For use with the Hematrak automated differential system
Quantity 16 oz.
Product Type Trak 300 Oil