Learn More
Epredia™ Richard-Allan™ Tissue-Loc™ TurbOflow™ Cassettes
Catalog No. 22045718 Shop All Epredia Products
This item is not returnable. View return policy
Catalog No. 22-045-718 Supplier Epredia™ Supplier No. 175480
This item is not returnable. View return policy
Uniquely designed to hold specimens securely and ensure optimal fluid exchange, up to 65% more than traditional cassettes
  • Helps reduce reagent carryover
  • When used with TurbOflow™ base molds, will use less paraffin and virtually eliminate flash
  • Constructed from durable, solvent resistant plastic