Description
Rotor; Thermo Scientific; For two shallow-well microplates; Holds two plates per run; RH2MP
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Type | Concentrator Rotor |
| For Use With (Equipment) | Savant ISS110 DNA SpeedVac Concentrator |
| Holds | 2 Plates |