Learn More
Thermo Scientific™ S1 Pipet Filler Accessories
GREENER_CHOICE
Catalog No. 14387044 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
14-387-044 1 g
1 options
Catalog No. 14-387-044 Supplier Thermo Scientific™ Supplier No. 9070
Add to Cart
Add to Cart
For use with S1 Pipet Filler
  • The wall-mounted holder and table stand offer compact storage between applications. It is possible to recharge the pipet filler in both stands by plugging in the charger. The wall-mounted holder is excellent for storing pipet filler when working in laminar flow cabinets to save space.
  • The wing stand† allows setting the S1 Pipet Filler down without removing or emptying the serological pipette.The easy-to-change filter prevents aerosol or liquid cross contamination of the pipette together with autoclavable silicone adapter. The 1 mL Pipette Support ensures that 1 mL pipettes will hold as securely as larger sizes.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Material Soda Lime Glass
Includes Dry glass microspheres
Quantity 1 g
† Not available for use or sale within the United States or for importation into the United States.