Learn More
Epredia™ Shandon™ AN-65-CH with Spreader Bars and Chains
Catalog No. 860090 Shop All Epredia Products
Change view
Click to view available options
Mechanism:
Manual
Catalog No. Mechanism
860090 Manual
1 options
Catalog No. 860090 Supplier Epredia™ Supplier No. 860090
  • Available as self-propelled or manual unit Uses chains and spreader bars to lift standard No. 82618 or No. 82619 body trays (sold separately; see page XXX); chains attach easily to body tray; also available with spreader bars and strap assembly upon request Designed to move body-laden trays to and from mortuary refrigerators with telescoping storage racks Well suited for use with side-opening refrigerators
Specifications
Mechanism Manual