Learn More
Epredia™ Shandon™ HEPA Bone Dust Collector Nozzle Adapters
Catalog No. 105461 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Bracket Extension
Nozzle Adapter
For Use With (Equipment):
Shandon 450 Autopsy Saw
Stryker Autopsy Saw
Stryker Autopsy Saw, Shandon 10000 Autopsy Saw
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Equipment)
105461 Nozzle Adapter Shandon 450 Autopsy Saw
10014 Bracket Extension Stryker Autopsy Saw
1000004 Nozzle Adapter Stryker Autopsy Saw, Shandon 10000 Autopsy Saw
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 105461 Supplier Epredia™ Supplier No. 105461
Requires
  • Lipshaw nozzle adapter: 105461 and 105468 necessary for complete assembly.
  • 10000/Stryker 90° nozzle adapters: 1000004 and 10015 necessary for complete assembly.
Fit HEPA Bone Dust Collector to an autopsy saw
  • Nozzle adapter (105462) for Stryker™ Autopsy Saw replaces standard HEPA Bone Dust Collector nozzle
  • Bracket extension (10014) for Stryker Autopsy Saw attaches standard nozzle to saw
Order Info
  • Shandon 450 Nozzle Adapter 105461 and 105468 necessary for complete assembly. Shandon 10000/Stryker 90° nozzle adapters 1000004 and 10015 necessary for complete assembly.
Specifications
Product Type Nozzle Adapter
For Use With (Equipment) Shandon 450 Autopsy Saw