Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon™ Mucolexx™

Catalog No. 9990371 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 x 3.8L (1 gal.)
4 x 500mL (0.13 gal.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9990371 4 x 3.8L (1 gal.)
9990370 4 x 500mL (0.13 gal.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 9990371 Supplier Epredia™ Supplier No. 9990371
Add to Cart
Add to Cart
This item is not returnable. View return policy

 

  • Ready-to-use mucoliquefying preservative
  • Liquefies (with agitation) mucoid cytology specimens
  • Maintains cellular detail
  • Short-term preservation of specimens

 

  • Histology
  • Cytology

Specifications

Color Light Red
Product Type Tissue Preservative
Quantity 4 x 3.8L (1 gal.)
For Use With (Application) Provides short term preservation of specimens
Form Liquid
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.