Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Shandon UniSystem™ Collection Pack

Catalog No. 200202 Shop All Epredia Products
Change view
Click to view available options
Volume (Metric):
90 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
20-020-2 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-020-2 Supplier Epredia™ Supplier No. 200202

Includes

  • 500 UniSystem Tubes
  • 500 Caps & Labels
  • 500 (90mL) Plastic Collection Cups
  • One Transport Rack

Contains everything necessary to collect 500 specimens

  • Produce consistent 0.4mL decanted urine sediment samples
  • Special design reduces inconsistencies in sampling and testing
  • The simple sampling technique minimizes the risk of exposureand cross-contamination

Specifications

Product Type UniSystem Collection Pack
Includes 500 UniSystem tubes, 500 caps and labels, Five hundred 90 mL Plastic collection cups, 1 transport rack
Volume (Metric) 90 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.