Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SUN-SRi™ 8-425 Screw Thread Caps

Catalog No. 14823304
Change view
Click to view available options
Color:
Black
Red
White
Yellow
Style:
Flanged
Not flanged
Quantity:
100/Pk.
1000/Cs.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Style Quantity
14-823-304 Red Not flanged 1000/Cs.
14-823-171 White Not flanged 1000/Cs.
14-823-323 Yellow Flanged 1000/Cs.
22-021869 Black Not flanged 100/Pk.
22-021871 Yellow Flanged 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-823-304 Supplier Thermo Scientific™ Supplier No. 500 016

Offered in a range of colors, and as open top, solid top or skirted Shimadzu™.

  • Polypropylene
  • For use with standard opening vials

TRUSTED_SUSTAINABILITY

Specifications

Color Red
Material Polypropylene
Style Not flanged
Quantity 1000/Cs.
Closure Size 8-425
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.