Learn More
Thermo Scientific™ SUN-SRi™ Hot Shot™ Guns
Catalog No. 14823464 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Cartridges (5 mL)
Catalog No. For Use With (Equipment)
14-823-464 Cartridges (5 mL)
1 options
Catalog No. 14-823-464 Supplier Thermo Scientific™ Supplier No. 59440HS
Generate three times greater force than manual filtration
The Hot Shot system is comprised of a solvent resistant cartridge pre-assembled with a TITAN filter containing a 3.9cm2 filtration area with a 1 micron, glass microfiber pre-filter increasing sample throughput. This process allows more sample to be processed before clogging causing frequent changing of filter units. Hot Shot«s built-in safety feature releases the cartridge before the pressure rating of the filter is exceeded, preventing bursting or “blow-off” of the filter.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hot Shot Gun
For Use With (Equipment) Cartridges (5 mL)