Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heracell™ Support Frame with Casters
GSA_VA

Catalog No. 13998053 Shop All Thermo Scientific Products
Catalog No. 13-998-053 Supplier Thermo Scientific™ Supplier No. 50057161
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Use With: Heracell 150i CO2 Incubator

  • Provide protection against floor contamination
  • Choice of three heights: 185mm (7.3 in.), 200mm (7.9 in.), or 780mm (30.7 in.)
  • Support frames can also be caster-mounted for easy maneuverability

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Heracell 150i CO2 incubator
Lighting LED
Type Support Frame
Includes Caster
Height (English) 7.3 in.
Height (Metric) 18.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.