Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SureTect™ Tools Starter Pack

Catalog No. A56898 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Application):
SureTect PCR Assay Workflow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
A56898 SureTect PCR Assay Workflow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A56898 Supplier Thermo Scientific™ Supplier No. A56898
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ease of sample setup and reduced handling steps enabled using the SureTect Starter Pack Tools.

Enhance the ease of use of each step of the SureTect workflow, promoting efficiency and productivity with Thermo Scientific™ SureTect™ Tools Starter Pack.

The Thermo Scientific SureTect Tools Starter Pack is a suite of tools specifically designed to enhance the ease of use of each step of the SureTect PCR Assay Workflow, promoting efficiency and productivity.

Consisting of everything needed to enhance the Thermo Scientific™ SureTect PCR Assay Workflow the starter packs consists of:

  • Thermo Scientific™ 96-well PCR Base - Specifically designed to perfectly fit the SureTect lysis and assay tubes providing stability during pipetting and enabling fast and easy sample setup.
  • Thermo Scientific ™ PCR Strip Cutting Tool - Enables fast and easy cutting of the SureTect Assay and lysis tube strips to enable flexible sample throughput.
  • Thermo Scientific™ Lysis Tube Uncapping Tool  - Fast and easy uncapping of the lysis tubes.
  • Thermo Scientific™ PCR Capping Tool for SureTect Lysis and Assay plates - Efficient capping in both the lysis step and the PCR step of the SureTect PCR Assay Workflow.
  • Applied Biosystems™ SimpliAmp™ 5 Strip Plate Tray - Specifically designed to quickly and easily setup and transfer the lysis tubes into the SimpliAmp instrument.
  • Applied Biosystems™ QuantStudio™ 5 Strip Plate Tray - Specifically designed to quickly and easily setup, transfer and run the PCR assay in the Quant Studio 5 instrument.
  • Applied Biosystems™ 7500 Fast 96W Strip Plate Adaptor - specifically designed to run the PCR SureTect tubes in the Applied Biosystems™ 7500 Fast instrument.
  • Applied Biosystems™ SimpliAmp™ Strip Plate Tray Cover - securely pipette from the lysis tubes and centrifuge the assay tubes with minimal hands on interaction.
  • Thermo Scientific™ Lysis Tube Caps, domed – An alternative to the pierceable seals for the maximum flexibility in sample processing.

Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.

We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.

TRUSTED_SUSTAINABILITY

Specifications

Description SureTect™ Tools Starter Pack
Content And Storage Room Temperature
For Use With (Application) SureTect PCR Assay Workflow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.