Learn More
Thermo Scientific™ Thermolyne™ Briskheat™ Heating Mat Accessories
Catalog No. 1146376 Shop All Fisherbrand Products
Change view
Click to view available options
Material:
Aluminum
Catalog No. Material
11-463-76 Aluminum
1 options
Catalog No. 11-463-76
For use with all Thermolyne/BriskHeat Heating Mats and Blankets (above). High-temperature aluminum tape conducts heat better as it is wider and thus secures more easily and firmly to larger vessels. Note: high-temperature aluminum tape are applied over or under the heating mat or blanket, running in the same direction. Tape cannot be reused.
Page 846, 1 May become permanently attached to vessel after application.
Specifications
Material Aluminum
Product Type High Temperature Tape