Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Thermo Scientific™ TipCycle Recycle Kit with Recycle PP#5 Pipette Tips, Tip Racks
GREENER_CHOICE

Catalog No. 01000587 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (English):
17.6 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
01-000-587 17.6 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-000-587 Supplier Thermo Scientific™ Supplier No. RECYCLEKIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ TipCycle Recycle Kit mail-back program provides a simple and effective solution for recycling PP#5 nonhazardous pipette tips and racks. Includes plastic liner, twist tie, instructions, waiver, prepaid freight label . Complete waiver, attach freight label and return for recycling.

Thermo Scientific™ TipCycle Recycle Kit mail-back program provides a simple and effective solution for recycling PP#5 nonhazardous pipette tips and racks. Kit includes plastic liner, twist tie, instructions, profile form for recyclable plastics (waiver), prepaid freight label. Place liner in box, fill with nonhazardous pipette tips and racks. Seal bag with twist tie, include waiver in box, seal box, apply prepaid freight label, then ship. Recycled plastic is converted into more sustainable, long-lasting, and affordable plastic lumber for pallets, retaining walls and landscaping products.

  • Thermo Scientific™ TipCycle Recycle Kit mail-back program provides a simple and effective solution for recycling PP#5 nonhazardous pipette tips and racks
  • For use with any ThermoFisher Scientific branded PP#5 pipette tips.
  • Complete TipCycle Pipette Tip Recycling Kit includes: box, instructions for return, a box liner and liner tie to help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, return freight label placed onto top of recycle package, then simply ship the recycle package
  • Box dimensions in inch: 17.75 x 14.75 x 15.5 in., 17.6 gal.
Note: TipCycle Stand Alone Recycle Kit does not include pipette tips. One case of TipCycle Recycle Kit contains 5 packs of individual recycle kits.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 66.5 L
Capacity (English) 17.6 gal.
Color Brown
Dimensions (L x W x H) 17.75 x 14.75 x 15.5 in.
Disposable Single-use
Sterility Non-sterile
Autoclavable Not Autoclavable
Includes Each kit includes cardboard box receptacle, plastic liner, twist tie
For Use With (Application) Intended use for recycling non-hazardous PP#5 pipette tips and pipette tip racks
Material Cardboard
Product Type Recycling box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.