Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TruScan™ RM Next Generation Handheld Raman Analyzer

Catalog No. 17720561
Change view
Click to view available options
Weight (Metric):
<0.9 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
17-720-561 <0.9 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-720-561 Supplier Thermo Scientific™ Supplier No. 8000039101

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Performs rapid material identification at the point-of-need within seconds to decrease sampling costs and increase inventory turns

The TruScan RM analyzer offers an unparalleled solution for rapid material identification.

  • Features highly selective Raman spectroscopy
  • Fast, point of need raw material identification (10-15 seconds)
  • Portable handheld and weighs less than 2 lbs (0.9kg)
  • Clear PASS or FAIL results enable user independence and require no secondary interpretation
  • Broad compound coverage includes cellulose based products
  • Fast and easy method development through assisted signature/reference acquisition
  • Intuitive workflow adapted to production environment through batch reports and ease of data input using barcode scanner
  • Greatly reduces analysis cost and overall overhead
  • Enhanced 21 CFR Part 11 and GMP compliance with biometric login, complex password options, and full audit trail features
  • Non-contact analysis through plastic bags, glass containers, blister packs, and clear gel caps greatly eliminates potential staff exposure to hazardous chemicals and highly potent active pharmaceutical ingredients (API) during inspection
  • Easy fleet management feature enable instrument and identification methods cloning
  • 24/7/365 Live customer support
  • Battery operated instrument (4 hours on a single charge)
  • Qualification documentation and standard operating procedures available

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance 21 CFR Part 11 and GMP Compliance
Weight (English) <2 lb.
Weight (Metric) <0.9 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.