Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Documention
Shop All Thermo Scientific ProductsDescription
- IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Specifications
| For Use With (Equipment) | ThermoScientific ULT Freezers |
| Description | Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, open door and power loss testing. |
| Product Type | IQ/OQ Documentation |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | IQ/OQ Documentation |