Learn More
Epredia™ Lab Vision™ UltraVision™ Quanto Mouse on Mouse HRP Detection System
Catalog No. TL125QHDM Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
Catalog No. Quantity
TL125QHDM 125 mL
1 options
Catalog No. TL125QHDM Supplier Epredia™ Supplier No. TL125QHDM
Add to Cart
Add to Cart
Request a Quote
HRP polymer designed to detect mouse antibodies on mouse tissue, maximizing signal while minimizing background.
Historically immunohistochemistry (IHC) staining with mouse antibodies on mouse tissue has been a challenge. The results were compromised by high levels of undesirable background or weak signal from interactions between the detection system and mouse tissues. The Epredia UltraVision Quanto Mouse on Mouse HRP Detection System eliminates these challenges, ensuring laboratories can produce optimal results routinely.
UltraVision Quanto Makes Optimal Results Routine by providing:
  • Strong Staining
  • Reduced Mouse on Mouse Background
  • Simple Biotin Free Polymer Procedure
  • For use with a DAB or other HRP Chromogen; For optimal results use with DAB Quanto TA-125-QHDX (125mL) or TA-060-QHDX (60mL)
Specifications
Quantity 125 mL