Learn More
Thermo Scientific™ VersaTREK™ Myco Media
GSA_VA
Catalog No. Y711142M Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
VersaTREK
Catalog No. For Use With (Equipment)
Y711142M VersaTREK
1 options
This item is not returnable. View return policy
Catalog No. Y711142M Supplier Thermo Scientific™ Supplier No. Y711142M
Add to Cart
Add to Cart
This item is not returnable. View return policy
Utilizes a unique growth matrix, via cellulose sponges, to provide better detection of all Mycobacteria species
VersaTREK Myco Media detects mycobacterial growth by automatically monitoring the rate of oxygen consumption within the headspace of the culture bottle every 24 minutes. A positive growth response is reported via a visible and audible positive signal by the system.
  • High bottle throughput, permits capacity of 4600 samples in a single VersaTREK 528 model
  • Comprehensive (FDA-cleared) full line of primary Mycobacterium tuberculosis antimicrobics
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) VersaTREK