Learn More
Thermo Scientific™ ViewPoint Enterprise Plug-Ins and Software
Catalog No. 19175218 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Viewpoint Enterprise Input Device Plug-Ins
Viewpoint Enterprise Plug-Ins
Viewpoint Enterprise Software
Description:
1 lot
Per 100 unit Package
Per 50 unit Package
Per unit
Third-party; Sold each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Description
19-175-218 Viewpoint Enterprise Input Device Plug-Ins Per unit
19-175-210 Viewpoint Enterprise Plug-Ins Third-party; Sold each
19-175-217 Viewpoint Enterprise Input Device Plug-Ins Per 50 unit Package
19-175-219 Viewpoint Enterprise Input Device Plug-Ins Per 100 unit Package
19-177-224 Viewpoint Enterprise Software 1 lot
19-177-225 Viewpoint Enterprise Software 1 lot
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-175-218 Supplier Thermo Scientific™ Supplier No. SYS/1/70853/015
Add to Cart
Add to Cart
Requires
System Specifications
Operates with Windows™ NT/2000/XP
Client Software Requirements
  • Microsoft Windows™ NT 4 Service Pack 6 or
  • Microsoft Windows™ 2000 Service Pack 4 or
  • Microsoft Windows™ XP Service Pack 1 or higher
Plug-ins and Software to be used with the ViewPoint Appliance. The platform is the base for a complete customizable solution for remote monitoring and remote real-time command and control; capable of integrating radiation, environmental, and general purpose detectors.
Plug-Ins
Each instrument, detector or sensor, which is capable of being remotely monitored by ViewPoint, has a unique Input Device Plug-In. This capability allows the instrument to communicate with ViewPoint Data Engine/System.
Client Plug-Ins
The ViewPoint User Interface and Data Engine are compatible with third-party Remote Monitoring Software systems. Compatability includes such systems as: MGPI-Teleview™, Industrial Innovative Solutions (IIS) – RADS™, and Bartlett – RMS™, and ESRI ArcView™ version 3.3 and 8. All purchased plug-ins are preloaded on the ViewPoint Appliance prior to delivery. Additional Input Device plug-ins, Client plug-ins, and User/Workstation License Bundles can be purchased and added to the original ViewPoint appliance via a patch file and updated security key
  • Real-time personnel dosimetry and data management
  • Area monitoring
  • Integrated remote monitoring
  • Instrument management
  • First Responder environmental monitoring
  • Site Process Monitoring
TRUSTED_SUSTAINABILITY
Specifications
Product Type Viewpoint Enterprise Input Device Plug-Ins
Description Per unit
Operating System Windows™ NT/2000/XP
For Use With (Application) Real time personnel dosimetry and data management, area monitoring, integrated remote monitoring, instrument management, first responder environmental monitoring, site process monitoring