Learn More
Unity Lab Services™ Thermo Scientific™ Water Purification Validation Documention for Thermo Scientific™ Barnstead LabTower EDI
Catalog No. 11675366 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Barnstead LabTower EDI
Catalog No. For Use With (Equipment)
11-675-366 Thermo Scientific Barnstead LabTower EDI
1 options
Catalog No. 11-675-366 Supplier Unity Lab Services™ Supplier No. IOQDOCE50133916
Add to Cart
Add to Cart
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Water Purification Validation Documention for Thermo Scientific Barnstead LabTower EDI
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Barnstead LabTower EDI
Description Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, Flow Adjustment, Water Pressure, Resistivity, Power Failure testing.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ Documentation