Learn More
Unity Lab Services™ Thermo Scientific™ Water Purification Validation Documention for Thermo Scientific™ Barnstead Smart2Pure, all models
Catalog No. 11675361 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Barnstead Smart2Pure, all models
Catalog No. For Use With (Equipment)
11-675-361 Thermo Scientific Barnstead Smart2Pure, all models
1 options
Catalog No. 11-675-361 Supplier Unity Lab Services™ Supplier No. IOQDOCE50133911
Add to Cart
Add to Cart
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Water Purification Validation Documention for Thermo Scientific Barnstead Smart2Pure, all models
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Barnstead Smart2Pure, all models
Description Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, Flow Adjustment, Water Pressure, Resistivity, Power Failure testing.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ Documentation