Learn More
Unity Lab Services™ Thermo Scientific™ Water Purification Validation Documention for Thermo Scientific™ Legacy Barnstead Reverse Osmosis
Shop All Thermo Scientific ProductsDescription
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Specifications
| For Use With (Equipment) | Thermo Scientific Legacy Barnstead Reverse Osmosis |
| Description | Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, Flow Adjustment, Water Pressure, Resistivity, Power Failure testing. |
| Location | Contact your local sales representative for a list of standard and extended coverage areas. |
| Includes | IQ/OQ Documentation |