Learn More
Thomas Scientific DTS-4 Digital Thermo Microplate Shaker for 4 Microplates, 100-240V; 50/60Hz
SDP
Catalog No. 501457546
Encompass
With the ability to accommodate either 2 or 4 microplates simultaneously, the DTS-2 and DTS-4 enable a considerable increase in processed volume. These high – precision space – savers free up valuable lab space while still ensuring a superior level of performance. 1.5 mm Rotational Orbit The DTS series features an ideal rotational orbit of 1.5mm for maximum performance, and works conveniently with any standard microplate. Bi – Directional Heating System The hermetic thermostatic container of each unit is equipped with a bi-directional heating system housed within both the base and the lid. Impervious to room temperature, this component allows the entire volume to be heated uniformly while eliminating condensation at the same time. The result – pinpoint accuracy and reliability with even micro-volume samples. Simultaneous Display of all Adjustive Parameters With a user-friendly display panel showing temperature, RPM, and timer settings separately, technicians can
Catalog No. 50-145-7546 Supplier Thomas Scientific Supplier No. 1185W28
With the ability to accommodate either 2 or 4 microplates simultaneously, the DTS-2 and DTS-4 enable a considerable increase in processed volume. These high – precision space – savers free up valuable lab space while still ensuring a superior level of performance. 1.5 mm Rotational Orbit The DTS series features an ideal rotational orbit of 1.5mm for maximum performance, and works conveniently with any standard microplate. Bi – Directional Heating System The hermetic thermostatic container of each unit is equipped with a bi-directional heating system housed within both the base and the lid. Impervious to room temperature, this component allows the entire volume to be heated uniformly while eliminating condensation at the same time. The result – pinpoint accuracy and reliability with even micro-volume samples. Simultaneous Display of all Adjustive Parameters With a user-friendly display panel showing temperature, RPM, and timer settings separately, technicians can