Learn More
Thermco Products Inc Block heater bottle thermometer, spirit, 18 deg./60 deg.C, 0.5C div, 125mm, 1ml mineral oil
Catalog No. NC0602749
Encompass_Preferred
We’re currently building out the product information and will be adding additional content soon. Thank you for your patience as we continue to expand our selection and improve your ordering experience.
Catalog No. NC0602749 Supplier Thermco Products Inc Supplier No. ACCDB301