Learn More
ATCC Thermoanaerobacterium; thermosaccharolyticum; Strain NCA 3814
Catalog No. 502384998
Encompass
Thermoanaerobacterium thermosaccharolyticum strain NCA 3814 is a thermophilic anaerobe. This bacterial type strain is whole-genome sequenced and can be propagated on PRAS-PYG with tween 80.
Catalog No. 50-238-4998 Supplier ATCC Supplier No. 7956
Thermoanaerobacterium thermosaccharolyticum strain NCA 3814 is a thermophilic anaerobe. This bacterial type strain is whole-genome sequenced and can be propagated on PRAS-PYG with tween 80.