Learn More
Cole-Parmer TSI 9545-A VelociCalc Air Velocity Meter; Articulating Probe/Auto Log
Catalog No. 50633409 Shop All Cole-Parmer Products
Encompass
These meters simultaneously measure and datalog several ventilation parameters using a single probe with multiple sensors. Great for cleanroom filtration airflow measurements they automatically calculate volumetric flow rate for round, square, and rectangular areas. All models measure velocity, temperature and calculate flow.
Catalog No. 50-633-409 Supplier Cole-Parmer Supplier No. 1027137
These meters simultaneously measure and datalog several ventilation parameters using a single probe with multiple sensors. Great for cleanroom filtration airflow measurements they automatically calculate volumetric flow rate for round, square, and rectangular areas. All models measure velocity, temperature and calculate flow.