Learn More
Cole-Parmer TSI 9535-A VelociCalc Air Velocity Meter; Articulating Probe/Manual Log
Catalog No. 50633407 Shop All Cole-Parmer Products
Encompass
These meters simultaneously measure and datalog several ventilation parameters using a single probe with multiple sensors. Great for cleanroom filtration airflow measurements — they automatically calculate volumetric flow rate for round, square, and rectangular areas. All models measure velocity, temperature and calculate flow.
Catalog No. 50-633-407 Supplier Cole-Parmer Supplier No. 1027135
These meters simultaneously measure and datalog several ventilation parameters using a single probe with multiple sensors. Great for cleanroom filtration airflow measurements — they automatically calculate volumetric flow rate for round, square, and rectangular areas. All models measure velocity, temperature and calculate flow.