Learn More
Cole-Parmer Extech 45158 Mini Thermoanemometer with Temperature and Humidity
Catalog No. 50992920 Shop All Cole-Parmer Products
Encompass
These anemometers feature a dual LCD to display air velocity with temperature, and windchill. Feature selectable average function, one-second display update, maximum reading, hold function, and auto shutoff after 20 minutes.
Catalog No. 50-992-920 Supplier Cole-Parmer Supplier No. 1020120
These anemometers feature a dual LCD to display air velocity with temperature, and windchill. Feature selectable average function, one-second display update, maximum reading, hold function, and auto shutoff after 20 minutes.