Learn More
Cole-Parmer Extech 45118 Mini Thermoanemometer with Temperature
Supplier:  Cole-Parmer 1020110
Encompass
These anemometers feature a dual LCD to display air velocity with temperature, and windchill. Feature selectable average function, one-second display update, maximum reading, hold function, and auto shutoff after 20 minutes.
Catalog No. 50-992-917