Learn More
Brookfield Engineer Labs THERMOCELL 115V
Catalog No. NC0278323
Encompass
  • Precise temperature control unit designed for accurate viscosity measurements.
  • Suitable for small sample volumes and elevated temperatures.
  • Stable thermal environment ideal for characterizing hot melts, asphalt, waxes, and polymers.
  • Programmable controller with single set points or up to 10 programmable temperature points.
  • Temperature ramping capabilities with integrated RheocalcT Software.
  • Solid-state proportional temperature controller and RTD probe for reliable data acquisition.
  • Compatible with standard Brookfield Viscometers and DV3T/DVNext Rheometers.
  • Requires external circulating water bath for cooling capability.
  • Includes control box, thermo container, RTD temperature probe, cooling rack, alignment bracket, extracting tool, pliers, and power cord.
  • Sample chamber made of stainless steel and includes disposable aluminum chambers.
Catalog No. NC0278323 Supplier Brookfield Engineer Labs Supplier No. HT115A DP
  • Precise temperature control unit designed for accurate viscosity measurements.
  • Suitable for small sample volumes and elevated temperatures.
  • Stable thermal environment ideal for characterizing hot melts, asphalt, waxes, and polymers.
  • Programmable controller with single set points or up to 10 programmable temperature points.
  • Temperature ramping capabilities with integrated RheocalcT Software.
  • Solid-state proportional temperature controller and RTD probe for reliable data acquisition.
  • Compatible with standard Brookfield Viscometers and DV3T/DVNext Rheometers.
  • Requires external circulating water bath for cooling capability.
  • Includes control box, thermo container, RTD temperature probe, cooling rack, alignment bracket, extracting tool, pliers, and power cord.
  • Sample chamber made of stainless steel and includes disposable aluminum chambers.