Learn More
Azenta US Inc Thermo-Conductive Tube Rack, Grey
Catalog No. NC3314383
Encompass
Thermo-Conductive Tube Rack, holds 6 strips wells and 12 x 1.5 or 2ml microcentrifuge tubes, 48 tapered wells for strips and 12 cylindrical wells, SBS compatible, grey
Catalog No. NC3314383 Supplier Azenta US Inc Supplier No. BCS523
Thermo-Conductive Tube Rack, holds 6 strips wells and 12 x 1.5 or 2ml microcentrifuge tubes, 48 tapered wells for strips and 12 cylindrical wells, SBS compatible, grey