Learn More
Chemglass Life Sciences Thermocouple, Type J, 0-370°C, 24 Guage, 1/8" OD, 6" Long Stainless Steel Probe, 48" Long Fiberglass Insulated Duplex Lead with Stainless Steel Overbraid for Liquid Immersion
SDP
Catalog No. CG1501801
Encompass_Preferred
Thermocouple, Type J, 0-370¿C, 24 Gauge, 1/8" OD, 6" Long Stainless Steel Probe, 48" Long Fiberglass Insulated Duplex Lead with Stainless Steel Overbraid for Liquid Immersion. Type ¿J¿ thermocouple (black) for use with oil baths or other liquid immersion applications. Thermocouple is supplied with a mini plug and a insulated and stainless steel overbraided cord. Thermocouples can be used to 370¿C.
Catalog No. CG1501801 Supplier Chemglass Life Sciences Supplier No. CG1501801
Thermocouple, Type J, 0-370¿C, 24 Gauge, 1/8" OD, 6" Long Stainless Steel Probe, 48" Long Fiberglass Insulated Duplex Lead with Stainless Steel Overbraid for Liquid Immersion. Type ¿J¿ thermocouple (black) for use with oil baths or other liquid immersion applications. Thermocouple is supplied with a mini plug and a insulated and stainless steel overbraided cord. Thermocouples can be used to 370¿C.