Learn More
Chemglass Life Sciences Clip, Thermocouple/Thermometer, 1/8 or 1/4" OD
SDP
Catalog No. CG110012
Encompass_Preferred
Clip, Thermocouple/Thermometer, 1/8 or 1/4" OD. Replacement part for CG1100 oil baths.
Catalog No. CG110012 Supplier Chemglass Life Sciences Supplier No. CG110012
Clip, Thermocouple/Thermometer, 1/8 or 1/4" OD. Replacement part for CG1100 oil baths.